Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromoanthraquinone 95.0+%, TCI America™
SDP

Catalog No. B26885G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26885G 5 g
B26881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B26885G Supplier TCI America Supplier No. B26885G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 632-83-7
Molecular Formula C14H7BrO2
Molecular Weight (g/mol) 287.112
InChI Key CXTPIHZYOGDSLV-UHFFFAOYSA-N
PubChem CID 12444
IUPAC Name 1-bromoanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)Br

Specifications

Melting Point 191°C
Quantity 5 g
Formula Weight 287.11
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromoanthraquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.