accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OClNOCH3N

CAS RN 6320-15-6

IUPAC Name: 4-chloro-2,6-dimethoxypyrimidine
Synonyms: 4-chloro-2,6-dimethoxypyrimidine
Molecular Weight (g/mol): 174.58
Molecular Formula: C6H7ClN2O2
InChi Key: JHNRTJRDRWKAIW-UHFFFAOYSA-N
SMILES: COC1=CC(Cl)=NC(OC)=N1
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)