accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6322-56-1

IUPAC Name: 4-acetyl-2-nitrobenzen-1-olate
Synonyms: 4-acetyl-2-nitrobenzenolate
Molecular Weight (g/mol): 180.14
Molecular Formula: C8H6NO4
InChi Key: MMNKVWGVSHRIJL-UHFFFAOYSA-M
SMILES: CC(=O)C1=CC=C([O-])C(=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results