Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Hydroxy-3'-nitroacetophenone 98.0+%, TCI America™
SDP

Catalog No. H09685G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H09685G Supplier TCI America Supplier No. H09685G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6322-56-1
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00017002
InChI Key MMNKVWGVSHRIJL-UHFFFAOYSA-N
Synonym 4'-hydroxy-3'-nitroacetophenone, 4-hydroxy-3-nitroacetophenone, 1-4-hydroxy-3-nitrophenyl ethanone, 4-acetyl-2-nitro-phenol, 1-4-hydroxy-3-nitrophenyl-1-ethanone, 1-4-hydroxy-3-nitrophenyl ethan-1-one, 2-nitro-4-acetylphenol, 4-acetyl-2-nitrophenol, 1-4-hydroxy-3-nitro-phenyl ethanone, ethanone, 1-4-hydroxy-3-nitrophenyl
PubChem CID 138723
SMILES CC(=O)C1=CC(=C(C=C1)O)[N+](=O)[O-]

Specifications

Melting Point 133°C
Quantity 5 g
Formula Weight 181.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Hydroxy-3′-nitroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.