accessibility menu, dialog, popup

UserName

Viewing profile as user:
SCNNO

CAS RN 63224-35-1

IUPAC Name: 4-(2-isothiocyanatoethyl)morpholine
Synonyms: 4-(2-isothiocyanatoethyl)morpholine
Molecular Weight (g/mol): 172.25
Molecular Formula: C7H12N2OS
InChi Key: SDNSCXCXVQHBGH-UHFFFAOYSA-N
SMILES: S=C=NCCN1CCOCC1
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)