CAS RN 6324-11-4
IUPAC Name:
2-(2-hydroxyphenoxy)acetic acid
Synonyms:
O-hydroxyphenoxyacetic acid
Molecular Weight (g/mol):
168.15
Molecular Formula:
C8H8O4
InChi Key:
LPXHPAYRONCQIF-UHFFFAOYSA-N
SMILES:
OC(=O)COC1=CC=CC=C1O
1
–
1
of
1
results
1