accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63245-28-3

IUPAC Name: 2-[(carboxymethyl)({[(2,6-diethylphenyl)carbamoyl]methyl})amino]acetic acid
Synonyms: [(carboxymethyl)({[(2,6-diethylphenyl)carbamoyl]methyl})amino]acetic acid
Molecular Weight (g/mol): 322.36
Molecular Formula: C16H22N2O5
InChi Key: WNIDXAKKFOKNEF-UHFFFAOYSA-N
SMILES: CCC1=CC=CC(CC)=C1NC(=O)CN(CC(O)=O)CC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results