Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2,6-Diethylphenylcarbamoylmethyl)iminodiacetic Acid 99.0+%, TCI America™
SDP

Catalog No. D17355G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D17355G Supplier TCI America Supplier No. D17355G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 63245-28-3
Molecular Formula C16H22N2O5
Molecular Weight (g/mol) 322.361
MDL Number MFCD00059631
InChI Key WNIDXAKKFOKNEF-UHFFFAOYSA-N
PubChem CID 170344
IUPAC Name 2-[carboxymethyl-[2-(2,6-diethylanilino)-2-oxoethyl]amino]acetic acid
SMILES CCC1=C(C(=CC=C1)CC)NC(=O)CN(CC(=O)O)CC(=O)O

Specifications

Melting Point 190°C
Color Red-White
Quantity 5 g
Formula Weight 322.36
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2,6-Diethylphenylcarbamoylmethyl)iminodiacetic Acid
RTECSNumber AG9712500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.