accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOHNSHN

CAS RN 6325-91-3

IUPAC Name: 5-nitro-2,3-dihydro-1H-1,3-benzodiazole-2-thione
Synonyms: 5-nitro-1,3-dihydro-1,3-benzodiazole-2-thione
Molecular Weight (g/mol): 195.2
Molecular Formula: C7H5N3O2S
InChi Key: YPXQSGWOGQPLQO-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2NC(=S)NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results