accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOCl

CAS RN 6334-96-9

IUPAC Name: 1-chloro-4-(4-chlorobutoxy)butane
Synonyms: butane, 1,1'-oxybis(4-chloro-
Molecular Weight (g/mol): 199.12
Molecular Formula: C8H16Cl2O
InChi Key: PVBMXMKIKMJQRK-UHFFFAOYSA-N
SMILES: ClCCCCOCCCCCl
Boiling Point 
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (4)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)