accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3CH

CAS RN 6336-58-9

IUPAC Name: dibutyl(prop-2-yn-1-yl)amine
Synonyms: dibutyl(prop-2-yn-1-yl)amine
Molecular Weight (g/mol): 167.3
Molecular Formula: C11H21N
InChi Key: XTEBRELWTGWYDE-UHFFFAOYSA-N
SMILES: CCCCN(CCCC)CC#C
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results