Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Dibutylamino-1-propyne 98.0+%, TCI America™
SDP

Catalog No. D28175ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28175ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D28175ML Supplier TCI America Supplier No. D28175ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Terminal alkyne reagent used for conjugation and click chemistry applications.

Chemical Identifiers

CAS 6336-58-9
Molecular Formula C11H21N
Molecular Weight (g/mol) 167.296
MDL Number MFCD00041673
InChI Key XTEBRELWTGWYDE-UHFFFAOYSA-N
Synonym 3-dibutylamino-1-propyne, dibutylprop-2-ynylamine, dibutylpropargylamine, 3-dibutylaminopropyne, n,n-dibutylpropargylamine, di-n-butyl propargyl amine, acmc-209nf4, di-n-butylamino-1-propyne, n,n-dibutyl-2-propynylamine
PubChem CID 138732
IUPAC Name N-butyl-N-prop-2-ynylbutan-1-amine
SMILES CCCCN(CCCC)CC#C

Specifications

Color Yellow
Quantity 5 mL
Formula Weight 167.30
Percent Purity ≥98.0% (T)
Grade Not Available
Physical Form Liquid
Chemical Name or Material 3-Dibutylamino-1-propyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.