accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 634-47-9

IUPAC Name: 2-chloro-4-methylquinoline
Synonyms: 2-chloro-4-methylquinoline
Molecular Weight (g/mol): 177.63
Molecular Formula: C10H8ClN
InChi Key: PFEIMKNQOIFKSW-UHFFFAOYSA-N
SMILES: CC1=C2C=CC=CC2=NC(Cl)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)