Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorolepidine 98.0+%, TCI America™
SDP

Catalog No. C01915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C01915G 5 g
C019125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C01915G Supplier TCI America Supplier No. C01915G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 634-47-9
Molecular Formula C10H8ClN
Molecular Weight (g/mol) 177.631
MDL Number MFCD00006742
InChI Key PFEIMKNQOIFKSW-UHFFFAOYSA-N
Synonym 2-chlorolepidine, 2-chloro-4-methyl-quinoline, quinoline, 2-chloro-4-methyl, lepidine, 2-chloro, pubchem5879, acmc-209nfp, 2-chloro-4methylquinoline, 4-methyl-2-chloroquinoline, 2-chlor-4-methyl-chinolin, 2-chloro-4-metyhylquinoline
PubChem CID 69449
IUPAC Name 2-chloro-4-methylquinoline
SMILES CC1=CC(=NC2=CC=CC=C12)Cl

Specifications

Melting Point 58°C
Color White-Yellow
Boiling Point 296°C
Quantity 5 g
Formula Weight 177.63
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chlorolepidine
EINECSNumber (5)-0720
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.