accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 634-90-2

IUPAC Name: 1,2,3,5-tetrachlorobenzene
Synonyms: 1,2,3,5-tetrachlorobenzene C6H2cl4
Molecular Weight (g/mol): 215.88
Molecular Formula: C6H2Cl4
InChi Key: QZYNWJQFTJXIRN-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(Cl)C(Cl)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results