Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,5-Tetrachlorobenzene 98.0+%, TCI America™
SDP

Catalog No. T08705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T08705G 5 g
T087025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T08705G Supplier TCI America Supplier No. T08705G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 634-90-2
Molecular Formula C6H2Cl4
Molecular Weight (g/mol) 215.88
MDL Number MFCD00000543
InChI Key QZYNWJQFTJXIRN-UHFFFAOYSA-N
Synonym benzene, 1,2,3,5-tetrachloro, unii-i27n186cin, ccris 5936, acmc-209nfx, dsstox_cid_6089, 1,3,5-tetrachlorobenzene, 1,2,3,5-tetrachlorbenzol, dsstox_rid_78013, dsstox_gsid_26089, ksc454c5l
PubChem CID 12468
ChEBI CHEBI:36696
IUPAC Name 1,2,3,5-tetrachlorobenzene
SMILES ClC1=CC(Cl)=C(Cl)C(Cl)=C1

Specifications

Melting Point 52°C
Color White-Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Boiling Point 246°C
Formula Weight 215.88
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1,2,3,5-Tetrachlorobenzene
EINECSNumber (3)-0076
RTECSNumber DB9445000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.