accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3NH2O

CAS RN 634-95-7

IUPAC Name: 1,1-diethylurea
Synonyms: N,N-diethylurea
Molecular Weight (g/mol): 116.16
Molecular Formula: C5H12N2O
InChi Key: TUMNHQRORINJKE-UHFFFAOYSA-N
SMILES: CCN(CC)C(N)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results