Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Diethylurea 98.0+%, TCI America™
SDP

Catalog No. D156725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D156725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D156725G Supplier TCI America Supplier No. D156725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 634-95-7
Molecular Formula C5H12N2O
Molecular Weight (g/mol) 116.164
MDL Number MFCD00007960
InChI Key TUMNHQRORINJKE-UHFFFAOYSA-N
Synonym n,n-diethylurea, asym-diethylurea, urea, n,n-diethyl, diethylurea, urea, 1,1-diethyl, urea, diethyl, urea,n-diethyl, urea,1-diethyl, 1,1-diethyl urea, acmc-1b3li
PubChem CID 12472
IUPAC Name 1,1-diethylurea
SMILES CCN(CC)C(=O)N

Specifications

Melting Point 68°C
Color White
Boiling Point 130°C
Quantity 25 g
Formula Weight 116.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Diethylurea
RTECSNumber YS9353900
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.