accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH2NNHNHOO

CAS RN 6341-24-8

IUPAC Name: 2-amino-3-(1H-imidazol-5-yl)propanoic acid hydrochloridyl
Synonyms: histidine hydrochloridyl
Molecular Weight (g/mol): 190.61
Molecular Formula: C6H9ClN3O2
InChi Key: IRHBVTLJVRUHBJ-UHFFFAOYNA-N
SMILES: [Cl].NC(CC1=CN=CN1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results