Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-histidine monohydrochloride hydrate, 99.2%, MP Biomedicals™

Catalog No. ICN10194925 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10194925 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10194925 Supplier MP Biomedicals, Inc Supplier No. 0210194925
Add to Cart
Add to Cart

Specifications

CAS 6341-24-8
Color White
Assay Percent Range 0.992
Molecular Formula C6H9ClN3O2
MDL Number MFCD00012702,MFCD00150081
Synonym d-histidine hydrochloride monohydrate, d-histidine monohydrochloride monohydrate, d-histidine hydrate hydrochloride, d-alpha-amino-beta-4-imidazolyl propionic acid monohydrochloride, d-histidine monohydrochloride monohydrate tlc, d-histidine monohydrochloride monohydrate at, d-2-degrees +/->>u-3-4-ssasso>>u +/-uea micro yennieani, r-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride monohydrate
InChI Key IRHBVTLJVRUHBJ-UHFFFAOYNA-N
SMILES [Cl].NC(CC1=CN=CN1)C(O)=O
IUPAC Name 2-amino-3-(1H-imidazol-5-yl)propanoic acid hydrochloridyl
Quantity 25 g
Molecular Weight (g/mol) 190.61
PubChem CID 15556510
Formula Weight 191.62
Physical Form Powder
Chemical Name or Material D-Histidine monohydrochloride hydrate
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.