accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6341-92-0

IUPAC Name: 6-chloro-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 6-chloro-1H-indole-2,3-dione
Molecular Weight (g/mol): 181.58
Molecular Formula: C8H4ClNO2
InChi Key: RVXLBLSGEPQBIO-UHFFFAOYSA-N
SMILES: ClC1=CC=C2C(NC(=O)C2=O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results