accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH2OCH3

CAS RN 63448-63-5

IUPAC Name: 1-methoxybutan-2-amine
Synonyms: 1-methoxybutan-2-amine
Molecular Weight (g/mol): 103.17
Molecular Formula: C5H13NO
InChi Key: QAFODUGVXFNLBE-UHFFFAOYNA-N
SMILES: CCC(N)COC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results