accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNNSOOHOSOOHOH2ONa

CAS RN 63451-29-6

IUPAC Name: 4-[3-(pyridin-2-yl)-5-(4-sulfophenyl)-1,2,4-triazin-6-yl]benzene-1-sulfonic acid hydrate sodium
Synonyms: ferrozine hydrate sodium
Molecular Weight (g/mol): 511.48
Molecular Formula: C20H16N4NaO7S2
InChi Key: TXAJQBKMPKPCAD-UHFFFAOYSA-N
SMILES: O.[Na].OS(=O)(=O)C1=CC=C(C=C1)C1=C(N=C(N=N1)C1=CC=CC=N1)C1=CC=C(C=C1)S(O)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results