Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(2-Pyridyl)-5,6-Diphenyl-1,2,4-Triazine-P,P'-Disulfonic Acid Monosodium Salt Hydrate

Catalog No. 1606015G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-060-15G 5 g
16-060-11G 1 g
16-060-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. 16-060-15G Supplier Sigma Aldrich Supplier No. 1606015G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent for the determination of Fe and Co, and Ag, Cu, and Ni.FerroZine is a trademark of Hach Company

Specifications

CAS 63451-29-6
Molecular Formula C20H13N4NaO6S2 -+ xH2O
Linear Formula C20H13N4NaO6S2 -+ xH2O
MDL Number MFCD00150794
Quantity 5 g
Synonym FerroZine(TM) Iron Reagent; PDT disulfonate monosodium salt hydrate
Molecular Weight (g/mol) 492.46 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.