accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6346-09-4

IUPAC Name: 4,4-diethoxybutan-1-amine
Synonyms: 4,4-diethoxy-butylamine
Molecular Weight (g/mol): 161.25
Molecular Formula: C8H19NO2
InChi Key: GFLPSABXBDCMCN-UHFFFAOYSA-N
SMILES: CCOC(CCCN)OCC
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (5)
  • (1)
  • (3)
  • (3)
  • (5)