accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3MgBr

CAS RN 63488-10-8

IUPAC Name: bromo(4-tert-butylphenyl)magnesium
Synonyms: bromo(4-tert-butylphenyl)magnesium
Molecular Weight (g/mol): 237.42
Molecular Formula: C10H13BrMg
InChi Key: YECFWFOEQLAVNH-UHFFFAOYSA-M
SMILES: CC(C)(C)C1=CC=C([Mg]Br)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)