accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2ClNOO

CAS RN 635-22-3

IUPAC Name: 4-chloro-3-nitroaniline
Synonyms: benzenamine, 4-chloro-3-nitro-
Molecular Weight (g/mol): 172.57
Molecular Formula: C6H5ClN2O2
InChi Key: FOHHWGVAOVDVLP-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C(=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results