Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-nitroaniline 99.0+%, TCI America™
SDP

Catalog No. C113825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C113825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C113825G Supplier TCI America Supplier No. C113825G
Add to Cart
Add to Cart

Specifications

CAS 635-22-3
Melting Point 105°C
Molecular Formula C6H5ClN2O2
Quantity 25 g
MDL Number MFCD00007844
UN Number 2237
Synonym benzenamine, 4-chloro-3-nitro, 3-nitro-4-chloroaniline, aniline, 4-chloro-3-nitro, unii-b13u77zl3a, 4-amino-1-chloro-2-nitrobenzene, 4-chloro-3-nitro aniline, 4-chloro-3-nitrophenylamine, pubchem21906, 3-nitro-4-chloraniline, wln: zr dg cnw
InChI Key FOHHWGVAOVDVLP-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1N)[N+](=O)[O-])Cl
IUPAC Name 4-chloro-3-nitroaniline
Molecular Weight (g/mol) 172.568
PubChem CID 12477
Formula Weight 172.57
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-nitroaniline
Show More Show Less
EINECSNumber (3)-0407
RTECSNumber BX1750000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.