accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 635-26-7

IUPAC Name: hydrogen (2-methylphenyl)hydrazine chloride
Synonyms: hydrogen (2-methylphenyl)hydrazine chloride
Molecular Weight (g/mol): 158.63
Molecular Formula: C7H11ClN2
InChi Key: KJGFNDCSTWGUDT-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC=CC=C1NN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results