Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Tolylhydrazine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. T03185G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T03185G 5 g
T031825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T03185G Supplier TCI America Supplier No. T03185G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 635-26-7
Molecular Formula C7H11ClN2
Molecular Weight (g/mol) 158.629
MDL Number MFCD00012925
InChI Key KJGFNDCSTWGUDT-UHFFFAOYSA-N
Synonym o-tolylhydrazine hydrochloride, 2-methylphenyl hydrazine hydrochloride, 2-methylphenylhydrazine hydrochloride, o-tolylhydrazine hcl, 1-2-methylphenyl hydrazine hydrochloride, 2-tolylhydrazine hydrochloride, o-tolylhydrazinium 1+ chloride, o-methylphenylhydrazine hcl, 2-methylphenylhydrazine hcl, hydrazine, 2-methylphenyl-, monohydrochloride
PubChem CID 71554
IUPAC Name (2-methylphenyl)hydrazine;hydrochloride
SMILES CC1=CC=CC=C1NN.Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 158.63
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material o-Tolylhydrazine Hydrochloride
EINECSNumber (3)-0468
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.