accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 635-39-2

IUPAC Name: 2-amino-6,7-dihydro-3H-purin-6-one hydrochloride
Synonyms: 2-aminohypoxanthine hydrochloride
Molecular Weight (g/mol): 187.59
Molecular Formula: C5H6ClN5O
InChi Key: IBAOFQIOOBQLHE-UHFFFAOYSA-N
SMILES: Cl.NC1=NC(=O)C2=C(N1)N=CN2
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)