Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanine Hydrochloride Hydrate 98.0+%, TCI America™
SDP

Catalog No. G017025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G017025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G017025G Supplier TCI America Supplier No. G017025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 635-39-2
Molecular Formula C5H6ClN5O
Molecular Weight (g/mol) 187.59
MDL Number MFCD00213670
InChI Key IBAOFQIOOBQLHE-UHFFFAOYSA-N
Synonym guanine hydrochloride, guanine hcl, 2-amino-1h-purin-6 7h-one hydrochloride, guanine, monohydrochloride, usaf s-1, guanine, hydrochloride, 6h-purin-6-one, 2-amino-1,7-dihydro-, monohydrochloride, 2-amino-9h-purin-6-ol hydrochloride 1:1, 6h-purin-6-one, 2-amino-1,9-dihydro-, hydrochloride 1:1, 2-amino-3,7-dihydropurin-6-one hydrochloride
PubChem CID 69459
IUPAC Name 2-amino-6,7-dihydro-3H-purin-6-one hydrochloride
SMILES Cl.NC1=NC(=O)C2=C(N1)N=CN2

Specifications

Color White
Quantity 25 g
Formula Weight 187.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Guanine Hydrochloride Hydrate
RTECSNumber MF8400000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.