accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2COOOCH2

CAS RN 63521-92-6

IUPAC Name: pent-4-enoyl pent-4-enoate
Synonyms: pent-4-enoyl pent-4-enoate
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: NEDHQDYBHYNBIF-UHFFFAOYSA-N
SMILES: C=CCCC(=O)OC(=O)CCC=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)