Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Pentenoic anhydride

Catalog No. 4718015ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-180-15ML 5 mL
47-180-125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 47-180-15ML Supplier Sigma Aldrich Supplier No. 4718015ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-pentenoic anhydride is a carboxylic anhydride.4-pentenoic anhydride may be used:
  • In the preparation of glucose functionalized (co)polymers
  • As a monomer in the preparation of cross-linked polyanhydrides
  • For the preparation of polymers with pendant vinyl or acetylene

Specifications

CAS 63521-92-6
Density 0.997 g/mL (at 25°C (literature))
Boiling Point 78°C to 81°C (0.4 mmHg)
Molecular Formula C10H14O3
Linear Formula (H2C=CHCH2CH2CO)2 O
Refractive Index n20/D 1.447 (literature)
MDL Number MFCD01320671
Quantity 5 mL
Molecular Weight (g/mol) 182.22
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.