accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63547-22-8

IUPAC Name: 2-[(diphenylmethyl)sulfanyl]acetate
Synonyms: 2-[(diphenylmethyl)sulfanyl]acetate
Molecular Weight (g/mol): 257.33
Molecular Formula: C15H13O2S
InChi Key: HTHFEDOFDBZPRX-UHFFFAOYSA-M
SMILES: [O-]C(=O)CSC(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results