accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6358-06-1

IUPAC Name: 5-amino-2-chlorophenol
Synonyms: 5-amino-2-chlorophenol
Molecular Weight (g/mol): 143.57
Molecular Formula: C6H6ClNO
InChi Key: JSCNCRWPXOTDDZ-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C(O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results