Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-chlorophenol 98.0+%, TCI America™
SDP

Catalog No. A19765G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19765G 5 g
A197625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19765G Supplier TCI America Supplier No. A19765G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6358-06-1
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD01707858
InChI Key JSCNCRWPXOTDDZ-UHFFFAOYSA-N
Synonym 2-chloro-5-aminophenol, 4-chloro-3-hydroxyaniline, phenol, 5-amino-2-chloro, 5-amino-2-chloro-phenol, 3-amino-6-chlorophenol, pubchem21902, 3-hydroxy-4-chloroaniline, 4-chloro-3-hydroxy aniline, acmc-1b64d, ksc494c4r
PubChem CID 80676
IUPAC Name 5-amino-2-chlorophenol
SMILES C1=CC(=C(C=C1N)O)Cl

Specifications

Melting Point 160°C
Quantity 5 g
Formula Weight 143.57
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-chlorophenol
RTECSNumber SJ5720000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.