accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2ClClCl

CAS RN 636-30-6

IUPAC Name: 2,4,5-trichloroaniline
Synonyms: 2,4,5-trichloroaniline
Molecular Weight (g/mol): 196.46
Molecular Formula: C6H4Cl3N
InChi Key: GUMCAKKKNKYFEB-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=C(Cl)C=C1Cl
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)