Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,4,5-Trichloroaniline

Catalog No. T54003100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T54003100G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T54003100G Supplier Sigma Aldrich Supplier No. T54003100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Synchronous Ar-F and Ar-Sn Bond Formation Through Fluorostannylation of ArynesContinuous-Flow Synthesis of Trimethylsilylphenyl Perfluorosulfonate Benzyne Precursors

Specifications

CAS 636-30-6
Melting Point 93°C to 95°C (lit.)
Boiling Point 270°C (lit.)
Molecular Formula C6H4Cl3N
Linear Formula Cl3C6H2NH2
MDL Number MFCD00007662
Quantity 1 g
Molecular Weight (g/mol) 196.46
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.