accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3OHO

CAS RN 636-44-2

IUPAC Name: 2,5-dimethylfuran-3-carboxylic acid
Synonyms: 2,5-dimethylfuran-3-carboxylic acid
Molecular Weight (g/mol): 140.14
Molecular Formula: C7H8O3
InChi Key: CNTHHNPBADVTRY-UHFFFAOYSA-N
SMILES: CC1=CC(C(O)=O)=C(C)O1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)