Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethyl-3-furancarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. D38635G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D38635G Supplier TCI America Supplier No. D38635G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 636-44-2
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00082181
InChI Key CNTHHNPBADVTRY-UHFFFAOYSA-N
Synonym 2,5-Dimethyl-3-furoic Acid
PubChem CID 69465
IUPAC Name 2,5-dimethylfuran-3-carboxylic acid
SMILES CC1=CC(=C(O1)C)C(=O)O

Specifications

Melting Point 137°C
Color White-Yellow
Quantity 5 g
Formula Weight 140.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dimethyl-3-furancarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.