accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 636-79-3

IUPAC Name: hydrogen pyridine-3-carboxylic acid chloride
Synonyms: hydrogen niacin chloride
Molecular Weight (g/mol): 159.57
Molecular Formula: C6H6ClNO2
InChi Key: IBTWUVRCFHJPKN-UHFFFAOYSA-N
SMILES: [H+].[Cl-].OC(=O)C1=CC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results