Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Picolinic Acid HCl, MP Biomedicals

Catalog No. ICN216820 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN216820 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN216820 Supplier MP Biomedicals, Inc Supplier No. 0521682010

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 636-79-3
Molecular Formula C6H6ClNO2
Molecular Weight (g/mol) 159.569
InChI Key IBTWUVRCFHJPKN-UHFFFAOYSA-N
Synonym niacin hydrochloride, nicotinic acid hydrochloride, 3-pyridinecarboxylic acid, hydrochloride, nicotinic acid, hydrochloride, pyridine-3-carboxylic acid hydrochloride, 3-pyridinecarboxylic acid, hydrochloride 1:1, 59-67-6 parent, niacin hcl, nicotinic acid hydrochloride 1:1, 3-pyridinecarboxylic acid hydrochloride
PubChem CID 71558
IUPAC Name pyridine-3-carboxylic acid;hydrochloride
SMILES C1=CC(=CN=C1)C(=O)O.Cl

Specifications

Quantity 10 g
Chemical Name or Material 2-Picolinic Acid HCL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.