accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 636-99-7

IUPAC Name: 1-(4-nitrophenyl)hydrazinium chloride
Synonyms: 1-(4-nitrophenyl)hydrazinium chloride
Molecular Weight (g/mol): 189.6
Molecular Formula: C6H8ClN3O2
InChi Key: ZWWXDCOPVYATOQ-UHFFFAOYSA-N
SMILES: [Cl-].N[NH2+]C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)