Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenylhydrazine Hydrochloride 99%, Thermo Scientific™

Catalog No. AC416031000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC416031000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC416031000 Supplier Thermo Scientific Chemicals Supplier No. 416031000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 636-99-7
Molecular Formula C6H8ClN3O2
Molecular Weight (g/mol) 189.599
InChI Key ZWWXDCOPVYATOQ-UHFFFAOYSA-N
Synonym 4-nitrophenylhydrazine hydrochloride, 4-nitrophenyl hydrazine hydrochloride, 4-nitrophenyhydrazine hydrochloride, 4-nitrophenylhydrazinehydrochloride, 4-nitrophenyl hydrazine, hcl, p-nitrophenylhydrazine hcl, nitrophenylhydrazine hydrochloride, hydrazine, 4-nitrophenyl-, monohydrochloride, p-nitrophenylhydrazine hydrochloride
PubChem CID 5463401
IUPAC Name (4-nitrophenyl)hydrazine;hydrochloride
SMILES C1=CC(=CC=C1NN)[N+](=O)[O-].Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 200°C to 204°C
Color Orange to Pink
Infrared Spectrum Authentic
Quantity 100 g
Formula Weight 189.6
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenylhydrazine hydrochloride, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.