accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CBr

CAS RN 63619-59-0

IUPAC Name: 4-bromo-4'-pentyl-1,1'-biphenyl
Synonyms: 4-bromo-4'-pentyl-1,1'-biphenyl
Molecular Weight (g/mol): 303.24
Molecular Formula: C17H19Br
InChi Key: VXJTWTJULLKDPY-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=C(C=C1)C1=CC=C(Br)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results