Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-4'-pentylbiphenyl 99.0+%, TCI America™
SDP

Catalog No. B13055G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B13055G Supplier TCI America Supplier No. B13055G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 63619-59-0
Molecular Formula C17H19Br
Molecular Weight (g/mol) 303.24
MDL Number MFCD00060106
InChI Key VXJTWTJULLKDPY-UHFFFAOYSA-N
Synonym 4-Amyl-4′C-bromobiphenyl
PubChem CID 618709
IUPAC Name 4-bromo-4'-pentyl-1,1'-biphenyl
SMILES CCCCCC1=CC=C(C=C1)C1=CC=C(Br)C=C1

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 5 g
Formula Weight 303.24
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-4′-pentylbiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.