accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63628-26-2

IUPAC Name: 2-methoxy-2-(naphthalen-1-yl)propanoic acid
Synonyms: 2-methoxy-2-(naphthalen-1-yl)propanoic acid
Molecular Weight (g/mol): 230.26
Molecular Formula: C14H14O3
InChi Key: YVWMPILNFZOQSZ-UHFFFAOYNA-N
SMILES: COC(C)(C(O)=O)C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results