accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONHOOOHO

CAS RN 63648-73-7

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}pentanedioic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}pentanedioic acid
Molecular Weight (g/mol): 281.26
Molecular Formula: C13H15NO6
InChi Key: PVFCXMDXBIEMQG-UHFFFAOYNA-N
SMILES: OC(=O)CCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)