Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyloxycarbonyl-D-glutamic acid, 95%

Catalog No. AAH6140122
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6140122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6140122 Supplier Thermo Scientific Chemicals Supplier No. H6140122
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 63648-73-7 | C13H15NO6 | 281.264 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63648-73-7
Molecular Formula C13H15NO6
Molecular Weight (g/mol) 281.264
MDL Number MFCD00063193
InChI Key PVFCXMDXBIEMQG-SNVBAGLBSA-N
Synonym z-d-glu-oh, z-d-glutamic acid, n-carbobenzoxy-d-glutamic acid, z-d-glu, n-cbz-d-glutamic acid, n-benzyloxycarbonyl-d-glutamic acid, r-2-benzyloxy carbonyl amino pentanedioic acid, cbz-glu-oh, 2r-2-benzyloxy carbonyl amino pentanedioic acid, cbz-d-glu
PubChem CID 1607568
IUPAC Name (2R)-2-(phenylmethoxycarbonylamino)pentanedioic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CCC(=O)O)C(=O)O

Specifications

Melting Point 119°C to 121°C
Quantity 100 g
Formula Weight 281.26
Percent Purity 95%
Chemical Name or Material N-Benzyloxycarbonyl-D-glutamic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.